2-Cyan-4-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Cyano-4-(trifluoromethyl)benzeneboronic acid
BB-8820-1G
BB-8820-250M
COBBBB-8820-1GEA
983
EUR
COBBBB-8820-1G
COBBBB-8820-250M
2-Cyan-4-(trifluormethyl)phenylboronsäure
2-Cyan-4-(trifluormethyl)phenylboronsäure
Formel:
C₈H₅BF₃NO₂ Molecular Weight: 214.94 g/mol |
MDL:
MFCD13195679 CAS-Nummer: 1218790-84-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...