4-(Methylthio)-2-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5134-5G BB-5134-1G
COBBBB-5134-5GEA 595 EUR
COBBBB-5134-5G COBBBB-5134-1G
4-(Methylthio)-2-(trifluormethyl)phenylboronsäure
4-(Methylthio)-2-(trifluormethyl)phenylboronsäure
Formel: C₈H₈BF₃O₂S
MDL: MFCD09972107
CAS-Nummer: 1072945-99-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR