5'-Fluor-2'-hydroxy-3'-nitroacetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(5-Fluoro-2-hydroxy-3-nitrophenyl)ethanone

012085-5G 012085-1G
FLUO012085-5GEA 369 EUR
FLUO012085-5G FLUO012085-1G
5'-Fluor-2'-hydroxy-3'-nitroacetophenon
5'-Fluor-2'-hydroxy-3'-nitroacetophenon
Formel: C₈H₆FNO₄
Molecular Weight: 199.136 g/mol
MDL: MFCD03094007
CAS-Nummer: 70978-39-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR