N-Cyclopropyl-2-nitro-4-(trifluormethyl)anilin

Lieferant: COMBI-BLOCKS

AN-1256-5G AN-1256-1G
COBBAN-1256-5GEA 0 EUR
COBBAN-1256-5G COBBAN-1256-1G
N-Cyclopropyl-2-nitro-4-(trifluormethyl)anilin
N-Cyclopropyl-2-nitro-4-(trifluormethyl)anilin
Formel: C₁₀H₉F₃N₂O₂
MDL: MFCD06149926
CAS-Nummer: 887351-41-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR