4-Nitrobenzylamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Nitrophenyl)methanamine hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

191434-25G 191434-5G
SIAL191434-25GEA 568 EUR
SIAL191434-25G SIAL191434-5G
4-Nitrobenzylamin Hydrochlorid, Sigma-Aldrich®
4-Nitrobenzylamin Hydrochlorid
Formel: C₇H₉ClN₂O₂
Molecular Weight: 188.62 g/mol
MDL: MFCD00012863
CAS-Nummer: 18600-42-5
EINECS: 242-441-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR