4-Hydroxybenzylamin Hydrobromid
Lieferant: FLUOROCHEM
Synonyme:
4-(Aminomethyl)phenol hydrobromide
008872-1G
008872-5G
FLUO008872-1GEA
55.4
EUR
FLUO008872-1G
FLUO008872-5G
4-Hydroxybenzylamin Hydrobromid
4-Hydroxybenzylamin Hydrobromid
Formel:
C₇H₁₀BrNO Molecular Weight: 204.0644 g/mol |
MDL:
MFCD02258769 CAS-Nummer: 90430-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...