3,4-Dihydroxybenzylamin Hydrobromid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
858781-250MG
858781-1G
858781-5G
SIAL858781-250MGEA
52.8
EUR
SIAL858781-250MG
SIAL858781-1G
SIAL858781-5G
3,4-Dihydroxybenzylamin Hydrobromid, Sigma-Aldrich®
3,4-Dihydroxybenzylamin Hydrobromid
Formel:
C₇H₁₀BrNO₂ |
MDL:
MFCD00012859 CAS-Nummer: 16290-26-9 EINECS: 240-382-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...