2-(Benzylthio)ethanol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Hydroxyethyl Benzyl Sulphide, 2-(Benzylthio)ethan-1-ol

L12274.06 L12274.14
L12274.06EA 16.9 EUR
L12274.06 L12274.14
2-(Benzylthio)ethanol ≥98%
2-(Benzylthio)ethanol

Purity: 98% CAS No.: 26524-88-9 Molecular Formula: C9H12OS Formula Weight: 168.25 Refractive Index: 1.5760 Boiling Point: 168-169°/18mm MDL No.: MFCD00014039

Formel: C₉H₁₂OS
Molecular Weight: 168.25 g/mol
Siedepunkt: 168…169 °C (18 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014039
CAS-Nummer: 26524-88-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR