3-Brom-2-(2-fluorbenzyloxy)-5-methylphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-8177-1G
BB-8177-250M
COBBBB-8177-1GEA
330
EUR
COBBBB-8177-1G
COBBBB-8177-250M
3-Brom-2-(2-fluorbenzyloxy)-5-methylphenylboronsäure
3-Brom-2-(2-fluorbenzyloxy)-5-methylphenylboronsäure
Formel:
C₁₄H₁₃BBrFO₃ |
MDL:
MFCD06798104 CAS-Nummer: 849062-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...