3-Brom-2-(4-fluorbenzyloxy)-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Bromo-2-((4-fluorobenzyl)oxy)-5-methylphenyl)boronic acid

BB-8054-1G BB-8054-250M
COBBBB-8054-1GEA 470 EUR
COBBBB-8054-1G COBBBB-8054-250M
3-Brom-2-(4-fluorbenzyloxy)-5-methylphenylboronsäure
3-Brom-2-(4-fluorbenzyloxy)-5-methylphenylboronsäure
Formel: C₁₄H₁₃BBrFO₃
Molecular Weight: 338,9700 g/mol
MDL: MFCD06411360
CAS-Nummer: 849062-41-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR