4-Benzyloxy-2-(trifluormethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Benzyloxy-2-trifluoromethylphenylboronic acid pinacol ester

PN-6061-1G PN-6061-250M
COBBPN-6061-1GEA 0 EUR
COBBPN-6061-1G COBBPN-6061-250M
4-Benzyloxy-2-(trifluormethyl)phenylboronsäurepinakolester
4-Benzyloxy-2-(trifluormethyl)phenylboronsäurepinakolester
Formel: C₂₀H₂₂BF₃O₃
MDL: MFCD13195761
CAS-Nummer: 1218790-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR