2-Benzyloxy-5-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-(Benzyloxy)-5-(trifluoromethyl)phenylboronic acid
BB-2806-1G
BB-2806-5G
COBBBB-2806-1GEA
149
EUR
COBBBB-2806-1G
COBBBB-2806-5G
2-Benzyloxy-5-(trifluormethyl)phenylboronsäure
2-Benzyloxy-5-(trifluormethyl)phenylboronsäure
Formel:
C₁₄H₁₂BF₃O₃ |
MDL:
MFCD06409944 CAS-Nummer: 612833-41-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...