4-Benzyloxy-2-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-(Benzyloxy)-2-(trifluoromethyl)phenylboronic acid, (4-(Benzyloxy)-2-(trifluoromethyl)phenyl)boronic acid
BB-6061-1G
BB-6061-250M
COBBBB-6061-1GEA
259
EUR
COBBBB-6061-1G
COBBBB-6061-250M
4-Benzyloxy-2-(trifluormethyl)phenylboronsäure
4-Benzyloxy-2-(trifluormethyl)phenylboronsäure
Formel:
C₁₄H₁₂BF₃O₃ Molecular Weight: 296.500 g/mol |
MDL:
MFCD13195652 CAS-Nummer: 1217501-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...