3-(4-Methoxybenzyloxy)-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(4-Methoxy)benzyloxy-5-trifluoromethylphenylboronic acid

BB-3490-1G BB-3490-5G
COBBBB-3490-1GEA 0 EUR
COBBBB-3490-1G COBBBB-3490-5G
3-(4-Methoxybenzyloxy)-5-(trifluormethyl)phenylboronsäure
3-(4-Methoxybenzyloxy)-5-(trifluormethyl)phenylboronsäure
Formel: C₁₅H₁₄BF₃O₄
MDL: MFCD08689490
CAS-Nummer: 913835-77-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR