2-Brom-4'-(trifluormethyl)acetophenon
Lieferant: BIONET RESEARCH
Synonyme:
4-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one
11X-0702.500
11X-0702.1
11X-0702.5
11X-0702.10
BIOT11X-0702.500EA
0
EUR
BIOT11X-0702.500
BIOT11X-0702.1
BIOT11X-0702.5
BIOT11X-0702.10
2-Brom-4'-(trifluormethyl)acetophenon
2-Brom-4'-(trifluormethyl)acetophenon
Formel:
C₉H₆BrF₃O Molecular Weight: 267.0400 g/mol |
MDL:
MFCD00126489 CAS-Nummer: 383-53-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...