2-Brom-4'-(trifluormethyl)acetophenon

Lieferant: MENAI

Synonyme: 4-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one

ST2495.1 ST2495.5
MENAST2495.1EA 0 EUR
MENAST2495.1 MENAST2495.5
2-Brom-4'-(trifluormethyl)acetophenon
2-Brom-4'-(trifluormethyl)acetophenon
Formel: C₉H₆BrF₃O
MDL: MFCD00126489
CAS-Nummer: 383-53-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR