2-Brom-4'-(trifluormethyl)acetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one

M-Clarity™ Qualitätsstufe = MQ100

669466-1G 669466-5G
SIAL669466-1GEA 27 EUR
SIAL669466-1G SIAL669466-5G
2-Brom-4'-(trifluormethyl)acetophenon, Sigma-Aldrich®
2-Brom-4'-(trifluormethyl)acetophenon
Formel: C₉H₆BrF₃O
MDL: MFCD00126489
CAS-Nummer: 383-53-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR