4-Brom-2,3-dimethylanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

OT-0392-5G OT-0392-1G
COBBOT-0392-5GEA 0 EUR
COBBOT-0392-5G COBBOT-0392-1G
4-Brom-2,3-dimethylanilin Hydrochlorid
4-Brom-2,3-dimethylanilin Hydrochlorid
Formel: C₈H₁₁BrClN
MDL: MFCD15143583
CAS-Nummer: 1215205-95-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR