4-Isopropyl-3-methylaniline hydrochloride

Lieferant: BIONET RESEARCH

Synonyme: 4-Isopropyl-m-toluidiniumchlorid

MS-5000.500 MS-5000.1 MS-5000.10 MS-5000.5
BIOTMS-5000.500EA 0 EUR
BIOTMS-5000.500 BIOTMS-5000.1 BIOTMS-5000.10 BIOTMS-5000.5
4-Isopropyl-3-methylaniline hydrochloride
4-Isopropyl-3-methylaniline hydrochloride
Formel: C₁₀H₁₆ClN
Molecular Weight: 185,70 g/mol
MDL: MFCD00156562
CAS-Nummer: 4534-11-6
EINECS: 224-876-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR