4-Biphenylboronsäurepinakolester
Lieferant: FLUOROCHEM
010849-10G
010849-1G
010849-5G
FLUO010849-10GEA
252
EUR
FLUO010849-10G
FLUO010849-1G
FLUO010849-5G
4-Biphenylboronsäurepinakolester
4-Biphenylboronsäurepinakolester
Formel:
C₁₈H₂₁BO₂ Molecular Weight: 280.17 g/mol |
MDL:
MFCD07368291 CAS-Nummer: 144432-80-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...