2-Biphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2234-5G BB-2234-1G
COBBBB-2234-5GEA 88.4 EUR
COBBBB-2234-5G COBBBB-2234-1G
2-Biphenylboronsäure
2-Biphenylboronsäure
Formel: C₁₂H₁₁BO₂
Molecular Weight: 198,03 g/mol
MDL: MFCD00136929
CAS-Nummer: 4688-76-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR