3-Biphenylboronsäure

Lieferant: Thermo Scientific

Synonyme: [1,1'-Biphenyl]-3-ylboronic acid

AC35888.250 AC35888.1
MAYBAC35888.250EA 0 EUR
MAYBAC35888.250 MAYBAC35888.1
3-Biphenylboronsäure
3-Biphenylboronsäure
Formel: C₁₂H₁₁BO₂
Molecular Weight: 198,03 g/mol
MDL: MFCD01318102
CAS-Nummer: 5122-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR