3-Biphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
[1,1'-Biphenyl]-3-ylboronic acid
BB-2235-25G
BB-2235-1G
BB-2235-5G
BB-2235-100G
COBBBB-2235-25GEA
247
EUR
COBBBB-2235-25G
COBBBB-2235-1G
COBBBB-2235-5G
COBBBB-2235-100G
3-Biphenylboronsäure
3-Biphenylboronsäure
Formel:
C₁₂H₁₁BO₂ Molecular Weight: 198,03 g/mol |
MDL:
MFCD01318102 CAS-Nummer: 5122-95-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...