4-Biphenylboronsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
[1,1'-Biphenyl]-4-ylboronic acid
B23703.03
B23703.06
B23703.14
B23703.03EA
33.6
EUR
B23703.03
B23703.06
B23703.14
4-Biphenylboronsäure ≥97%
4-Biphenylboronsäure
Purity: 98+%
CAS No.: 5122-94-1
Molecular Formula: C12H11BO2
Formula Weight: 198.03
Odour: Odourless
Melting Point: 238-247°
Beilstein Reference: 2937410
MDL No.: MFCD00093311
Formel:
C₁₂H₁₁BO₂ Molecular Weight: 198,03 g/mol Schmelzpunkt: 238…247 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00093311 CAS-Nummer: 5122-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...