4-Biphenylboronsäure

Lieferant: CHEMPUR

Synonyme: [1,1'-Biphenyl]-4-ylboronic acid

SB02091.250 SB02091.1
CHMPSB02091.250EA 0 EUR
CHMPSB02091.250 MAYBSB02091.1
4-Biphenylboronsäure
4-Biphenylboronsäure
Formel: C₁₂H₁₁BO₂
Molecular Weight: 198,03 g/mol
MDL: MFCD00093311
CAS-Nummer: 5122-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR