4-Biphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: [1,1'-Biphenyl]-4-ylboronic acid

M-Clarity™ Qualitätsstufe = MQ200

483451-25G 483451-5G
SIAL483451-25GEA 87.9 EUR
SIAL483451-25G SIAL483451-5G
4-Biphenylboronsäure, Sigma-Aldrich®
4-Biphenylboronsäure
Formel: C₁₂H₁₁BO₂
Molecular Weight: 198,03 g/mol
MDL: MFCD00093311
CAS-Nummer: 5122-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR