2-Fluor-3-biphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Fluoro-[1,1'-biphenyl]-3-yl)boronic acid

BB-6208-5G BB-6208-1G BB-6208-250M
COBBBB-6208-5GEA 2040 EUR
COBBBB-6208-5G COBBBB-6208-1G COBBBB-6208-250M
2-Fluor-3-biphenylboronsäure
2-Fluor-3-biphenylboronsäure
Formel: C₁₂H₁₀BFO₂
Molecular Weight: 216,200 g/mol
MDL: MFCD15143460
CAS-Nummer: 1218790-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR