2-Fluor-4-biphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-4-biphenylylboronic acid, (3-fluoro-4-phenyl)phenylboronic acid, 3-Fluoro-4-phenylbenzeneboronic acid, 2-Fluoro-1,1'-biphenyl-4-ylboronic acid

BB-2723-5G BB-2723-25G BB-2723-1G
COBBBB-2723-5GEA 66.7 EUR
COBBBB-2723-5G COBBBB-2723-25G COBBBB-2723-1G
2-Fluor-4-biphenylboronsäure
2-Fluor-4-biphenylboronsäure
Formel: C₁₂H₁₀BFO₂
Molecular Weight: 216,02 g/mol
MDL: MFCD01075707
CAS-Nummer: 178305-99-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR