3-Benzylcarbamoyl-4-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Benzylcarbamoyl)-4-fluorophenylboronic acid

BB-2865-5G BB-2865-1G
COBBBB-2865-5GEA 1040 EUR
COBBBB-2865-5G COBBBB-2865-1G
3-Benzylcarbamoyl-4-fluorphenylboronsäure
3-Benzylcarbamoyl-4-fluorphenylboronsäure
Formel: C₁₄H₁₃BFNO₃
MDL: MFCD08235041
CAS-Nummer: 874219-22-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR