3-Fluor-4-(4-methoxybenzyloxycarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3409-5G BB-3409-1G
COBBBB-3409-5GEA 0 EUR
COBBBB-3409-5G COBBBB-3409-1G
3-Fluor-4-(4-methoxybenzyloxycarbamoyl)phenylboronsäure
3-Fluor-4-(4-methoxybenzyloxycarbamoyl)phenylboronsäure
Formel: C₁₅H₁₅BFNO₅
MDL: MFCD08235097
CAS-Nummer: 913835-49-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR