7-Chlor-4-hydroxy-8-methylchinolin-3-carbonsäureethylester

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 7-chloro-4-hydroxy-8-methylquinoline-3-carboxylate

HC-6382-5G HC-6382-1G
COBBHC-6382-5GEA 595 EUR
COBBHC-6382-5G COBBHC-6382-1G
7-Chlor-4-hydroxy-8-methylchinolin-3-carbonsäureethylester
7-Chlor-4-hydroxy-8-methylchinolin-3-carbonsäureethylester
Formel: C₁₃H₁₂ClNO₃
Molecular Weight: 265,6923 g/mol
MDL: MFCD00173377
CAS-Nummer: 5350-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR