7-Chlor-4-hydroxy-8-methylchinolin-3-carbonsäure

Lieferant: COMBI-BLOCKS

PY-7802-5G PY-7802-1G
COBBPY-7802-5GEA 564 EUR
COBBPY-7802-5G COBBPY-7802-1G
7-Chlor-4-hydroxy-8-methylchinolin-3-carbonsäure
7-Chlor-4-hydroxy-8-methylchinolin-3-carbonsäure
Formel: C₁₁H₈ClNO₃
MDL: MFCD02179783
CAS-Nummer: 405923-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR