7-Chlor-8-methylchinolin-4-ol
Lieferant: COMBI-BLOCKS
Synonyme:
7-Chloro-4-hydroxy-8-methylquinoline
HC-6478-5G
HC-6478-25G
HC-6478-1G
COBBHC-6478-5GEA
303
EUR
COBBHC-6478-5G
COBBHC-6478-25G
COBBHC-6478-1G
7-Chlor-8-methylchinolin-4-ol
7-Chlor-8-methylchinolin-4-ol
Formel:
C₁₀H₈ClNO |
MDL:
MFCD00272440 CAS-Nummer: 203626-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...