Kaliumselenocyanat, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ200
216186-10G
216186-250G
216186-50G
SIAL216186-10GEA
102
EUR
SIAL216186-10G
SIAL216186-250G
SIAL216186-50G
Kaliumselenocyanat, Sigma-Aldrich®
Potassium selenocyanate
Formel:
CKNSe |
MDL:
MFCD00011366 CAS-Nummer: 3425-46-5 EINECS: 222-320-1 UN: 3283 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...