Kaliumselenocyanat, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

483699-25G 483699-5G
SIAL483699-25GEA 140 EUR
SIAL483699-25G SIAL483699-5G
Kaliumselenocyanat, Sigma-Aldrich®
Potassium selenocyanate
Formel: CKNSe
Molecular Weight: 144.08 g/mol
MDL: MFCD00011366
CAS-Nummer: 3425-46-5
EINECS: 222-320-1
UN: 3283
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR