Siliciumnitrid
Lieferant: CHEMPUR SILANE
SILS14700.25
SILS14700.100
SILNSILS14700.25EA
0
EUR
SILNSILS14700.25
SILNSILS14700.100
Siliciumnitrid
Siliciumnitrid
Formel:
N₄Si₃ |
MDL:
MFCD00011230 CAS-Nummer: 12033-89-5 EINECS: 234-796-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...