Methyltetrahydropyran-4-carboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl tetrahydro-2H-pyran-4-carboxylate
AM-1008-5G
AM-1008-100G
AM-1008-25G
COBBAM-1008-5GEA
46
EUR
COBBAM-1008-5G
COBBAM-1008-100G
COBBAM-1008-25G
Methyltetrahydropyran-4-carboxylat
Methyltetrahydropyran-4-carboxylat
Formel:
C₇H₁₂O₃ Molecular Weight: 144.1684 g/mol |
MDL:
MFCD01075170 CAS-Nummer: 110238-91-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...