Methyltetrahydropyran-4-carboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl tetrahydro-2H-pyran-4-carboxylate

40199-250ML 40199-50ML
SIAL40199-250MLEA 546 EUR
SIAL40199-250ML SIAL40199-50ML
Methyltetrahydropyran-4-carboxylat, Sigma-Aldrich®
Methyltetrahydropyran-4-carboxylat
Formel: C₇H₁₂O₃
Molecular Weight: 144,1684 g/mol
MDL: MFCD01075170
CAS-Nummer: 110238-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR