4-Aminotetrahydropyran Hydrochlorid
Lieferant: COMBI-BLOCKS
Synonyme:
Tetrahydro-2H-pyran-4-amine hydrochloride
AM-1014-5G
AM-1014-1G
COBBAM-1014-5GEA
63.9
EUR
COBBAM-1014-5G
COBBAM-1014-1G
4-Aminotetrahydropyran Hydrochlorid
4-Aminotetrahydropyran Hydrochlorid
Formel:
C₅H₁₂ClNO Molecular Weight: 137,6079 g/mol |
MDL:
MFCD00100881 CAS-Nummer: 33024-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...