2-Pyridinsulfonsäure

Lieferant: COMBI-BLOCKS

Synonyme: Pyridine-2-sulphonic acid

PY-7844-1G PY-7844-25G PY-7844-5G
COBBPY-7844-1GEA 42.4 EUR
COBBPY-7844-1G COBBPY-7844-25G COBBPY-7844-5G
2-Pyridinsulfonsäure
2-Pyridinesulphonic acid
Formel: C₅H₅NO₃S
Molecular Weight: 159.1631 g/mol
MDL: MFCD00234080
CAS-Nummer: 15103-48-7
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR