3-Pyridinsulfonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pyridine-3-sulphonic acid

82820-100G 82820-25G
SIAL82820-100GEA 105 EUR
SIAL82820-100G SIAL82820-25G
3-Pyridinsulfonsäure, Sigma-Aldrich®
3-Pyridinesulphonic acid
Formel: C₅H₅NO₃S
Molecular Weight: 159.16 g/mol
MDL: MFCD00006381
CAS-Nummer: 636-73-7
EINECS: 211-265-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR