Strontium-2-ethylhexanoat 40% (8 - 12% Sr) in 2-Ethylhexansäure
Lieferant: Thermo Scientific
Achtung
Synonyme: 2-Ethylhexanoic acid strontium salt
40719.14 40719.22
40719.14EA 152 EUR
40719.14 40719.22
Strontium-2-ethylhexanoat 40% (8 - 12% Sr) in 2-Ethylhexansäure
Strontium-2-ethylhexanoat
CAS No.: 2457-02-5 Molecular Formula: C16H30O4Sr Formula Weight: 374.04 Physical Form: Liquid Odour: Mild MDL No.: MFCD00070438
Formel: C₁₆H₃₀O₄Sr
Molecular Weight: 374,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00070438
CAS-Nummer: 2457-02-5
EINECS: 219-536-3