Allyl-ethylsulfid ≥97%

Lieferant: Thermo Scientific
Gefahr

L12847.03 L12847.06
L12847.03EA 26.2 EUR
L12847.03 L12847.06
Allyl-ethylsulfid ≥97%
Allyl-ethylsulfid

Purity: 97% CAS No.: 5296-62-8 Molecular Formula: C5H10S Formula Weight: 102.19 Flash Point: 18°(64°F) Refractive Index: 1.4680 Odour: Unpleasant Boiling Point: 115-116° Beilstein Reference: 1736876 UN No.: 1993 MDL No.: MFCD00026979

Formel: C₅H₁₀S
Molecular Weight: 102,19 g/mol
Siedepunkt: 115…116 °C
Dichte: 0,868
Flammpunkt: 18 °C (64 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00026979
CAS-Nummer: 5296-62-8
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR