1-Nonansulfonsäure Natriumsalz, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Sodium 1-nonanesulphonate, Sodium nonane-1-sulphonate

M-Clarity™ Qualitätsstufe = MQ100

N0893-25G N0893-5G
SIALN0893-25GEA 1000 EUR
SIALN0893-25G SIALN0893-5G
1-Nonansulfonsäure Natriumsalz, Sigma-Aldrich®
1-Nonanesulphonic acid sodium salt
Formel: C₉H₁₉NaO₃S
Molecular Weight: 230,30 g/mol
MDL: MFCD00025010
CAS-Nummer: 35192-74-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR