Nickelsulfat Hydrat ≥99,9985% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
|
Gefahr
|
10820.14
10820.22
10820.14EA
112
EUR
10820.14
10820.22
Nickelsulfat Hydrat ≥99,9985% (Metall-Basis), Puratronic®
Nickelsulfat Hydrat
Purity: 99.9985%
CAS No.: 15244-37-8
Molecular Formula: NiO4S.xH2O (x≈6)
Formula Weight: 154.77(anhy)
Physical Form: Crystalline powder
Density: 2.070
Appearance: Green
Odour: Odourless
pH: 4.3-4.7
Merck Reference: 14,6517
UN No.: 3288
MDL No.: MFCD00011146
Formel:
NiO₄S·H₂O Molecular Weight: 154,77 g/mol (anhy) Dichte: 2,070 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011146 CAS-Nummer: 15244-37-8 EINECS: 232-104-9 UN: 3288 ADR: 6.1,III Merck Index: 14,6517 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...