3-Isopropoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Isopropoxybenzeneboronic acid

BB-2098-1G BB-2098-100G BB-2098-25G BB-2098-5G
COBBBB-2098-1GEA 33 EUR
COBBBB-2098-1G COBBBB-2098-100G COBBBB-2098-25G COBBBB-2098-5G
3-Isopropoxyphenylboronsäure
3-Isopropoxyphenylboronsäure
Formel: C₉H₁₃BO₃
Molecular Weight: 180,01 g/mol
MDL: MFCD03701512
CAS-Nummer: 216485-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR