4-Isopropoxyphenylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Isopropoxybenzeneboronic acid

H52675.03 H52675.06 H52675.14
H52675.03EA 33.3 EUR
H52675.03 H52675.06 H52675.14
4-Isopropoxyphenylboronsäure ≥97%
4-Isopropoxyphenylboronsäure

Purity: 97% CAS No.: 153624-46-5 Molecular Formula: C9H13BO3 Formula Weight: 180.01 Melting Point: 150-154° MDL No.: MFCD03427051

Formel: C₉H₁₃BO₃
Molecular Weight: 180,01 g/mol
Schmelzpunkt: 150…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03427051
CAS-Nummer: 153624-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR