4-Isopropoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Isopropoxybenzeneboronic acid

BB-2104-5G BB-2104-25G BB-2104-1G BB-2104-100G
COBBBB-2104-5GEA 26.2 EUR
COBBBB-2104-5G COBBBB-2104-25G COBBBB-2104-1G COBBBB-2104-100G
4-Isopropoxyphenylboronsäure
4-Isopropoxyphenylboronsäure
Formel: C₉H₁₃BO₃
Molecular Weight: 180,01 g/mol
MDL: MFCD03427051
CAS-Nummer: 153624-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR