4-(Octyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-n-Octyloxyphenylboronic acid, (4-(Octyloxy)phenyl)boronic acid, 4-n-Octyloxybenzeneboronic acid

BB-2737-5G BB-2737-25G BB-2737-1G
COBBBB-2737-5GEA 0 EUR
COBBBB-2737-5G COBBBB-2737-25G COBBBB-2737-1G
4-(Octyloxy)phenylboronsäure
4-(Octyloxy)phenylboronsäure
Formel: C₁₄H₂₃BO₃
Molecular Weight: 250.1400 g/mol
MDL: MFCD00982004
CAS-Nummer: 121554-09-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR