2-Fluor-5-(trifluormethoxy)benzylbromid ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

H26795.03 H26795.06
H26795.03EA 102 EUR
H26795.03 H26795.06
2-Fluor-5-(trifluormethoxy)benzylbromid ≥97%
2-Fluor-5-(trifluormethoxy)benzylbromid

Purity: 97% CAS No.: 86256-24-8 Molecular Formula: C8H5BrF4O Formula Weight: 273.02 Refractive Index: 1.4660

Formel: C₈H₅BrF₄O
Molecular Weight: 273.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115890
CAS-Nummer: 86256-24-8
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR